Assay Details
| Gene Symbol: IL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195219.1 | NP_001182148.1 | 2 - 3 | 490 | 64 | |
| XM_012180592.2 | XP_012035982.1 | 3 - 4 | 447 | 64 | ||
| GenBank mRNA | FR670343.1 | - | 2 - 3 | 490 | 64 | |
| FR670344.1 | - | 3 - 4 | 495 | 64 | ||
More Information
Set Membership: |
|

IL25
Genomic Map