Assay Details
| Gene Symbol: PRDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280704.1 | NP_001267633.1 | 1 - 2 | 191 | 59 | |
| GenBank mRNA | KC342239.1 | - | 1 - 2 | 191 | 59 | |
More Information
Set Membership: |
|

PRDX6
Genomic Map