Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308564.1 | NP_001295493.1 | 2 - 3 | 625 | 62 | |
| GenBank mRNA | DQ192015.1 | - | 2 - 3 | 381 | 62 | |
| KF925831.1 | - | 2 - 3 | 625 | 62 | ||
More Information
Set Membership: |
|

BMP7
Genomic Map