Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078660.1 | NP_001072128.1 | 3 - 4 | 201 | 64 | |
XM_027977374.1 | XP_027833175.1 | 4 - 5 | 347 | 64 | ||
XM_027977375.1 | XP_027833176.1 | 4 - 5 | 373 | 64 | ||
XM_027977376.1 | XP_027833177.1 | 4 - 5 | 347 | 64 | ||
GenBank mRNA | DQ496115.1 | - | 3 - 4 | 201 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
