Assay Details
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093786.1 | NP_001087255.1 | 2 | 691 | 73 | |
| XM_015091592.2 | XP_014947078.2 | 5 - 6 | 799 | 73 | ||
| XM_015091593.2 | XP_014947079.2 | 4 - 5 | 703 | 73 | ||
| GenBank mRNA | EF488163.1 | - | 2 | 691 | 73 | |
More Information
Set Membership: |
|

YBX1
Genomic Map