Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161872.1 | NP_001155344.1 | 2 | 517 | 81 | |
| GenBank mRNA | FJ943980.1 | - | 2 | 517 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASD1| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161872.1 | NP_001155344.1 | 2 | 517 | 81 | |
| GenBank mRNA | FJ943980.1 | - | 2 | 517 | 81 | |
Set Membership: |
|