Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187218.3 | XP_012042608.1 | 2 - 3 | 192 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP1LC3C| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187218.3 | XP_012042608.1 | 2 - 3 | 192 | 111 | |
Set Membership: |
|