Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190204.3 | XP_012045594.1 | 2 - 3 | 165 | 80 | |
| XM_012190205.3 | XP_012045595.1 | 2 - 3 | 202 | 80 | ||
| XM_012190206.3 | XP_012045596.1 | 1 - 2 | 1001 | 80 | ||
More Information
Set Membership: |
|

HCST
Genomic Map