Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098042.2 | XP_014953528.2 | 2 - 3 | 3025 | 71 | |
| XM_027973885.1 | XP_027829686.1 | 2 - 3 | 2731 | 71 | ||
More Information
Set Membership: |
|

PCDH8
Genomic Map