Assay Details
Gene Symbol: MAP3K8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014274.3 | XP_004014323.1 | 6 - 7 | 1166 | 67 | |
XM_015099629.2 | XP_014955115.1 | 6 - 7 | 1268 | 67 | ||
XM_027976700.1 | XP_027832501.1 | 6 - 7 | 4259 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
