Assay Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969448.1 | XP_027825249.1 | 4 - 5 | 666 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MVB12A| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969448.1 | XP_027825249.1 | 4 - 5 | 666 | 61 | |
Set Membership: |
|