Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006520.4 | XP_004006569.1 | 5 - 6 | 707 | 65 | |
| XM_004006521.4 | XP_004006570.1 | 5 - 6 | 637 | 65 | ||
More Information
Set Membership: |
|

TSPAN31
Genomic Map