Assay Details
Gene Symbol: PLAAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096872.2 | XP_011952262.2 | 4 - 5 | 705 | 77 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PLAAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096872.2 | XP_011952262.2 | 4 - 5 | 705 | 77 |