Assay Details
Gene Symbol: C1QTNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017061.4 | XP_004017110.1 | 5 - 6 | 753 | 70 | |
XM_004017062.4 | XP_004017111.1 | 5 - 6 | 981 | 70 | ||
XM_012097112.3 | XP_011952502.1 | 5 - 6 | 941 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
