Assay Details
| Gene Symbol: CCDC120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963398.1 | XP_027819199.1 | 4 - 5 | 2962 | 56 | |
| XM_027963399.1 | XP_027819200.1 | 5 - 6 | 1231 | 56 | ||
| XM_027963400.1 | XP_027819201.1 | 5 - 6 | 681 | 56 | ||
| XM_027963401.1 | XP_027819202.1 | 5 - 6 | 1584 | 56 | ||
| XM_027963402.1 | XP_027819203.1 | 4 - 5 | 421 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963401.1 transcripts.
Set Membership: |
|

CCDC120
Genomic Map