Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970294.1 | XP_027826095.1 | 3 - 4 | 429 | 75 | |
| XM_027970295.1 | XP_027826096.1 | 3 - 4 | 429 | 75 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map