Assay Details
| Gene Symbol: ANKRD54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968092.1 | XP_027823893.1 | 3 - 4 | 573 | 53 | |
| XM_027968093.1 | XP_027823894.1 | 3 - 4 | 208 | 53 | ||
More Information
Set Membership: |
|

ANKRD54
Genomic Map