Assay Details
| Gene Symbol: SPRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966690.1 | XP_027822491.1 | 3 - 4 | 1960 | 67 | |
| XM_027966691.1 | XP_027822492.1 | 3 - 4 | 1969 | 67 | ||
More Information
Set Membership: |
|

SPRED2
Genomic Map