Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966021.1 | XP_027821822.1 | 3 - 4 | 354 | 63 | |
| XM_027966022.1 | XP_027821823.1 | 3 - 4 | 440 | 63 | ||
More Information
Set Membership: |
|

ARRDC1
Genomic Map