Assay Details
| Gene Symbol: SNTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018347.4 | XP_004018396.3 | 2 - 3 | 210 | 73 | |
| XM_027958205.1 | XP_027814006.1 | 2 - 3 | 207 | 73 | ||
More Information
Set Membership: |
|

SNTN
Genomic Map