Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020888.4 | XP_004020937.1 | 4 - 5 | 745 | 61 | |
| XM_012104368.3 | XP_011959758.1 | 3 - 4 | 1779 | 61 | ||
| XM_012104369.3 | XP_011959759.1 | 4 - 5 | 732 | 61 | ||
| XM_012104371.3 | XP_011959761.1 | 4 - 5 | 1130 | 61 | ||
| XM_012104373.3 | XP_011959763.1 | 4 - 5 | 1084 | 61 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map