Assay Details
| Gene Symbol: LOC101121522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971442.1 | XP_027827243.1 | 11 - 12 | 1661 | 100 | |
| XM_027971443.1 | XP_027827244.1 | 11 - 12 | 1621 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101121522| Gene Symbol: LOC101121522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971442.1 | XP_027827243.1 | 11 - 12 | 1661 | 100 | |
| XM_027971443.1 | XP_027827244.1 | 11 - 12 | 1621 | 100 | ||