Assay Details
| Gene Symbol: TMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974180.1 | XP_027829981.1 | 6 - 7 | 1116 | 55 | |
| XM_027974181.1 | XP_027829982.1 | 6 - 7 | 1164 | 55 | ||
| XM_027974182.1 | XP_027829983.1 | 5 - 6 | 1286 | 55 | ||
| XM_027974183.1 | XP_027829984.1 | 6 - 7 | 1068 | 55 | ||
| XM_027974184.1 | XP_027829985.1 | 6 - 7 | 1118 | 55 | ||
| XM_027974185.1 | XP_027829986.1 | 6 - 7 | 1116 | 55 | ||
| XM_027974186.1 | XP_027829987.1 | 6 - 7 | 1114 | 55 | ||
| XM_027974187.1 | XP_027829988.1 | 6 - 7 | 1116 | 55 | ||
More Information
Set Membership: |
|

TMC8
Genomic Map