Assay Details
| Gene Symbol: KBTBD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957864.1 | XP_027813665.1 | 2 - 3 | 1257 | 67 | |
| XM_027957865.1 | XP_027813666.1 | 2 - 3 | 1290 | 67 | ||
| XM_027957866.1 | XP_027813667.1 | 1 - 2 | 1409 | 67 | ||
| XM_027957867.1 | XP_027813668.1 | 2 - 3 | 1223 | 67 | ||
More Information
Set Membership: |
|

KBTBD12
Genomic Map