Assay Details
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004923.3 | XP_004004972.1 | 6 - 7 | 881 | 68 | |
XM_012146099.2 | XP_012001489.1 | 6 - 7 | 905 | 68 | ||
XM_027965178.1 | XP_027820979.1 | 6 - 7 | 1238 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
