Assay Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007000.4 | XP_004007049.2 | 4 - 5 | 703 | 83 | |
| XM_027968129.1 | XP_027823930.1 | 3 - 4 | 5448 | 83 | ||
| XM_027968130.1 | XP_027823931.1 | 4 - 5 | 755 | 83 | ||
More Information
Set Membership: |
|

MIEF1
Genomic Map