Assay Details
| Gene Symbol: DNAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010777.3 | XP_004010826.1 | 3 - 4 | 221 | 81 | |
| XM_012182021.3 | XP_012037411.1 | 3 - 4 | 211 | 81 | ||
More Information
Set Membership: |
|

DNAL1
Genomic Map