Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961556.1 | XP_027817357.1 | 4 - 5 | 2428 | 56 | |
| XM_027961558.1 | XP_027817359.1 | 4 - 5 | 2148 | 56 | ||
More Information
Set Membership: |
|

NYAP1
Genomic Map