Assay Details
| Gene Symbol: NYAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961556.1 | XP_027817357.1 | 5 - 6 | 2649 | 87 | |
| XM_027961558.1 | XP_027817359.1 | 5 - 6 | 2369 | 87 | ||
| XM_027961559.1 | XP_027817360.1 | 5 - 6 | 2646 | 87 | ||
More Information
Set Membership: |
|

NYAP1
Genomic Map