Assay Details
| Gene Symbol: MAP7D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012106377.3 | XP_011961767.1 | 14 - 15 | 1867 | 69 | |
| XM_012106379.3 | XP_011961769.2 | 13 - 14 | 1682 | 69 | ||
| XM_012106381.3 | XP_011961771.1 | 13 - 14 | 1829 | 69 | ||
| XM_012106383.3 | XP_011961773.2 | 12 - 13 | 1571 | 69 | ||
| XM_012106384.3 | XP_011961774.1 | 14 - 15 | 1867 | 69 | ||
| XM_015104825.2 | XP_014960311.1 | 14 - 15 | 1805 | 69 | ||
| XM_015104826.2 | XP_014960312.1 | 14 - 15 | 1837 | 69 | ||
| XM_027963068.1 | XP_027818869.1 | 14 - 15 | 1748 | 69 | ||
More Information
Set Membership: |
|

MAP7D2
Genomic Map