Assay Details
| Gene Symbol: DCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010170.4 | XP_004010219.1 | 7 - 8 | 959 | 61 | |
| XM_004010171.4 | XP_004010220.1 | 7 - 8 | 959 | 61 | ||
More Information
Set Membership: |
|

DCP2
Genomic Map