Assay Details
| Gene Symbol: AGRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100491.2 | XP_014955977.2 | 2 - 3 | 936 | 91 | |
| GenBank mRNA | AY310396.1 | - | 2 - 3 | 129 | 91 | |
More Information
Set Membership: |
|

AGRP
Genomic Map