Assay Details
Gene Symbol: KRT40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012877.4 | XP_004012926.2 | 4 - 5 | 846 | 62 | |
GenBank mRNA | HQ283084.1 | - | 4 - 5 | 845 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
