Assay Details
| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020578.4 | XP_004020627.2 | 3 - 4 | 339 | 100 | |
| XM_027960812.1 | XP_027816613.1 | 2 - 3 | 285 | 100 | ||
More Information
Set Membership: |
|

SERPINB7
Genomic Map