Assay Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022505.4 | XP_004022554.1 | 2 - 3 | 348 | 77 | |
| GenBank mRNA | EF630627.1 | - | 1 - 2 | 147 | 77 | |
More Information
Set Membership: |
|

TNMD
Genomic Map