Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969735.1 | XP_027825536.1 | 5 - 6 | 1618 | 66 | |
| XM_027969736.1 | XP_027825537.1 | 5 - 6 | 1746 | 66 | ||
More Information
Set Membership: |
|

SNAPC2
Genomic Map