Assay Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014030.4 | XP_004014079.3 | 4 - 5 | 720 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
