Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190329.2 | XP_012045719.2 | 3 - 4 | 672 | 89 | |
| XM_027977402.1 | XP_027833203.1 | 3 - 4 | 1075 | 89 | ||
| XM_027977403.1 | XP_027833204.1 | 1 - 2 | 507 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027977403.1 transcripts.
Set Membership: |
|

ZNF527
Genomic Map