Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190329.2 | XP_012045719.2 | 4 - 5 | 796 | 88 | |
| XM_027977402.1 | XP_027833203.1 | 4 - 5 | 1199 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF527| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190329.2 | XP_012045719.2 | 4 - 5 | 796 | 88 | |
| XM_027977402.1 | XP_027833203.1 | 4 - 5 | 1199 | 88 | ||