Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958449.1 | XP_027814250.1 | 3 - 4 | 792 | 78 | |
| XM_027958450.1 | XP_027814251.1 | 3 - 4 | 736 | 78 | ||
More Information
Set Membership: |
|

FAM83B
Genomic Map