Assay Details
| Gene Symbol: ZNF652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974685.1 | XP_027830486.1 | 4 - 5 | 5703 | 72 | |
| XM_027974686.1 | XP_027830487.1 | 4 - 5 | 1490 | 72 | ||
More Information
Set Membership: |
|

ZNF652
Genomic Map