Assay Details
| Gene Symbol: LAYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979185.1 | XP_027834986.1 | 2 - 3 | 542 | 60 | |
| XM_027979186.1 | XP_027834987.1 | 2 - 3 | 542 | 60 | ||
| XM_027979187.1 | XP_027834988.1 | 2 - 3 | 542 | 60 | ||
| XM_027979188.1 | XP_027834989.1 | 2 - 3 | 542 | 60 | ||
| XM_027979189.1 | XP_027834990.1 | 2 - 3 | 542 | 60 | ||
More Information
Set Membership: |
|

LAYN
Genomic Map