Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107015.3 | XP_011962405.1 | 3 - 4 | 2262 | 76 | |
| XM_012107017.3 | XP_011962407.1 | 5 - 6 | 1738 | 76 | ||
| XM_027962818.1 | XP_027818619.1 | 5 - 6 | 1648 | 76 | ||
| XM_027962819.1 | XP_027818620.1 | 5 - 6 | 1089 | 76 | ||
| XM_027962820.1 | XP_027818621.1 | 6 - 7 | 1346 | 76 | ||
| XM_027962821.1 | XP_027818622.1 | 6 - 7 | 1351 | 76 | ||
| XM_027962822.1 | XP_027818623.1 | 4 - 5 | 983 | 76 | ||
| XM_027962823.1 | XP_027818624.1 | 5 - 6 | 1720 | 76 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map