Assay Details
| Gene Symbol: PRDM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097262.2 | XP_014952748.2 | 1 - 2 | 151 | 50 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM13| Gene Symbol: PRDM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097262.2 | XP_014952748.2 | 1 - 2 | 151 | 50 | |
Set Membership: |
|