Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956452.1 | XP_027812253.1 | 2 - 3 | 295 | 76 | |
| XM_027956453.1 | XP_027812254.1 | 2 - 3 | 593 | 76 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map