Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979963.1 | XP_027835764.1 | 4 - 5 | 725 | 81 | |
| XM_027979968.1 | XP_027835769.1 | 4 - 5 | 702 | 81 | ||
More Information
Set Membership: |
|

GTPBP8
Genomic Map