Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959995.1 | XP_027815796.1 | 3 - 4 | 381 | 68 | |
| XM_027959996.1 | XP_027815797.1 | 3 - 4 | 381 | 68 | ||
More Information
Set Membership: |
|

HRAS
Genomic Map