Assay Details
| Gene Symbol: SHISA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102442.2 | XP_014957928.2 | 5 - 6 | 658 | 107 | |
| XM_027957680.1 | XP_027813481.1 | 3 - 4 | 957 | 107 | ||
More Information
Set Membership: |
|

SHISA5
Genomic Map