Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962469.1 | XP_027818270.1 | 6 - 7 | 2512 | 76 | |
| XM_027962470.1 | XP_027818271.1 | 3 - 4 | 2469 | 76 | ||
More Information
Set Membership: |
|

MMRN2
Genomic Map