Assay Details
| Gene Symbol: MMP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977120.1 | XP_027832921.1 | 7 - 8 | 1306 | 66 | |
| XM_027977121.1 | XP_027832922.1 | 7 - 8 | 1180 | 66 | ||
More Information
Set Membership: |
|

MMP24
Genomic Map